MICROLOOP 10UL PL RED 20BAG 1 * 1.000 items
Catalog # MEDWMW71020
Supplier: MEDICAL WIRE EQUIPMENT
New Product
MICROLOOP 10UL PL RED 20BAG 1 * 1.000 items
Catalog # MEDWMW71020
Supplier: MEDICAL WIRE EQUIPMENT
New Product
Frequently Bought Together
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Disposable cuvettes
Supplier: Brand
VWR® Reagent Reservoirs
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Inoculating loops, disposable, Microloops
Supplier: MEDICAL WIRE EQUIPMENT