ContactUsLinkComponent
Long path rectangular cell holder for 20-100 mm pathlength cuvettes for Orion AquaMate 7100 & 8100 Spectrophotometers 1 * 1 items
Catalog # ORIOAQX1LWLVH
Supplier:  Thermo Orion
New Product
Long path rectangular cell holder for 20-100 mm pathlength cuvettes for Orion AquaMate 7100 & 8100 Spectrophotometers 1 * 1 items
Catalog # ORIOAQX1LWLVH
Supplier:  Thermo Orion