ContactUsLinkComponent
Aerotest SimultanTest CO2, complete kit with Tubes and impactor includes: box of 10 Dräger-Tubes nitrous gases (NOX) 0.2/a box of 10 Dräger-Tubes ammonia (NH3) 2/a box of 10 Dräger-Tubes phosphine (PH3) 0.1/c box of 10 Dräger-Tubes carbon monoxide (C 1 * 1 KIT
Catalog # DRAE6526170
Supplier:  Dräger
New Product
Aerotest SimultanTest CO2, complete kit with Tubes and impactor includes: box of 10 Dräger-Tubes nitrous gases (NOX) 0.2/a box of 10 Dräger-Tubes ammonia (NH3) 2/a box of 10 Dräger-Tubes phosphine (PH3) 0.1/c box of 10 Dräger-Tubes carbon monoxide (C 1 * 1 KIT
Catalog # DRAE6526170
Supplier:  Dräger