ContactUsLinkComponent
S3l hro fo sr ,high shoe nubuk brown/orange smellstop lining fresh'n flex puncture resistant fabric insole dry'n air omnia footb ed aluminium toe-cap double density pu/rubber outsole i-daptive system 1 * 1 PAIR
Catalog # PRTWB1042BKY39
Supplier:  Portwest
New Product
S3l hro fo sr ,high shoe nubuk brown/orange smellstop lining fresh'n flex puncture resistant fabric insole dry'n air omnia footb ed aluminium toe-cap double density pu/rubber outsole i-daptive system 1 * 1 PAIR
Catalog # PRTWB1042BKY39
Supplier:  Portwest