ContactUsLinkComponent
S1pl fo sr ankle shoe black/red water resistant technical textile and suedeleather smellstop lining fresh'n flex puncture resist ant fabric insole slimcap toe-cap mono pu outsole 1 * 1 PAIR
Catalog # PRTWB0983BBKD44
Supplier:  Portwest
New Product
S1pl fo sr ankle shoe black/red water resistant technical textile and suedeleather smellstop lining fresh'n flex puncture resist ant fabric insole slimcap toe-cap mono pu outsole 1 * 1 PAIR
Catalog # PRTWB0983BBKD44
Supplier:  Portwest