ContactUsLinkComponent
Muffle furnace with scale and software l9/11/sw/c550 tmax1100 with flap door and over-temperature limiter and thermocouple hole in t he furnace door 1 * 1 items
Catalog # NABELW095K1ANOTY
Supplier:  NABERTHERM
New Product
Muffle furnace with scale and software l9/11/sw/c550 tmax1100 with flap door and over-temperature limiter and thermocouple hole in t he furnace door 1 * 1 items
Catalog # NABELW095K1ANOTY
Supplier:  NABERTHERM