ContactUsLinkComponent
Muffle furnace with scale and software lt9/11/sw/b510 tmax1100 with lift door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # NABELW095H1ANLTY
Supplier:  NABERTHERM
New Product
Muffle furnace with scale and software lt9/11/sw/b510 tmax1100 with lift door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # NABELW095H1ANLTY
Supplier:  NABERTHERM