Order Entry
Belgium
Orders LinkContactUsLinkComponent
WHEATON® Rubber Stoppers

WHEATON® Rubber Stoppers

Stopper, natural red rubber, 13,0 mm, 20,0 mm
Catalog # 548-2050
Supplier:   DWK Life Sciences
WHEATON® Rubber Stoppers
Catalog # 548-2050
Supplier:  DWK Life Sciences
WHEATON® Rubber Stoppers
WHEATON® Rubber Stoppers

Stopper, natural red rubber, 13,0 mm, 20,0 mm

Price
699,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Stoppers of various styles, sizes and rubber formulations.

  • Manufactured from the highest quality raw materials
  • Components manufactured to yield exceptional dimensional stability
  • Autoclavable

Straight plug stoppers provide maximum sealing for vials with straight-wall glass finish.

Snap-on style provides maximum sealing for vials with a blow-back glass finish.

Sleeve style stoppers feature a fold-over skirt for sealing in serum finish vials without a crimp closure.

Specifications
  • Ø bottom:
    13,0 mm
  • Ø top:
    20,0 mm
  • Description:
    Stopper, natural red rubber
  • Seq:
    05
  • Cat. No.:
    548-2050
  • Durometer hardness:
    39
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.