High Stackable Ledges, 100 mm
Shelf ledge, Metroseal grey with Microban®, 457 mm width
Catalog # METOL18N-4K4
Supplier: Metro International
Aller à :
Product Details & Documents
Actual ledge length is approximately 25 mm shorter than nominal shelf length/width.
Spécifications
- Description:Shelf ledge, Metroseal grey with Microban®, 457 mm width
- Seq:06
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



