Orders LinkContactUsLinkComponent
Slide Trays
Slide Trays
Slide tray, White, PS, 194×297×12 mm, 20
Catalog # DELT989921
Supplier:   DELTALAB
Slide Trays
Catalog # DELT989921
Supplier:  DELTALAB
Slide Trays
Slide Trays
Slide tray, White, PS, 194×297×12 mm, 20
Price
106,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stackable slide trays of polystyrene.
Documents
Specifications
  • No. of slides:
    20
  • Colour:
    White
  • Description:
    Slide tray
  • Material:
    PS
  • Seq:
    02
  • W×D×H:
    194×297×12 mm