Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer Capsules

Buffer Capsules

pH buffer capsules, GPH 7, amount: 5 capsules, 7,0, Green, 1 SET
Catalog # GRSG602616
Supplier:   Greisinger
Buffer Capsules
Catalog # GRSG602616
Supplier:  Greisinger
CAS Number:  
Buffer Capsules

pH buffer capsules, GPH 7, amount: 5 capsules, 7,0, Green, 1 SET

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 SET
Product Details & Documents

Cost-effective alternative: Buffer capsules and working sets. With buffer capsules, you can produce a 100 ml buffer solution in the most cost-effective manner. A particular advantage of the capsules is that their storage life is several years longer than that of ready-to-use solutions.

The capsules are offered in a practical working set: The most cost-effective entry with everything you need for a successful start in pH measurement, including cleaning solution and KCL100 storage/replacement electrolyte.

Specifications

Specifications

  • Pk:
    1 SET
  • Colour:
    Green
  • Description:
    pH buffer capsules, GPH 7, amount: 5 capsules
  • pH value:
    7,0

Specifications