ContactUsLinkComponent
Microscope slides, 12x12+0/-0,4 mm, thickness 1 mm, white glass, cut edges, without frosted end, in boxes of 50 pcs., 500 pcs./carton, paper-interleaved, CE marked, MARIENFELD SUPERIOR 1 * 1 items
Catalog # MARI11300900927
Supplier:  MARIENFELD
New Product
Microscope slides, 12x12+0/-0,4 mm, thickness 1 mm, white glass, cut edges, without frosted end, in boxes of 50 pcs., 500 pcs./carton, paper-interleaved, CE marked, MARIENFELD SUPERIOR 1 * 1 items
Catalog # MARI11300900927
Supplier:  MARIENFELD