Prototypic, selective kainate receptor agonist. Potent excitant and neurotoxin. 1 * 25 mg
Catalog # MSPPHB0355-25MG
Supplier: MARKET SOURCE PART PROCESS
Prototypic, selective kainate receptor agonist. Potent excitant and neurotoxin. 1 * 25 mg
Catalog # MSPPHB0355-25MG
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Petri dishes, Corning® Gosselin™
Supplier: Corning
VIAL 55ML CUSTODY CLEAR 19 1 * 450 items
Supplier: Capitol Europe
Cell culture plates, 1536-well, CELLSTAR®
Supplier: Greiner Bio-One
60 mm Gridded Scoring Dish 1 * 20 items
Supplier: STEMCELL Technologies
Fibronectin-coated flasks, Corning® BioCoat®
Supplier: Corning
Microplates, 96-well
Supplier: KARTELL
Analytical 24-WELL PLATE ADAPTERS FOR DHTC 1 * 1 items
Supplier: MARKET SOURCE PART PROCESS
Storage cryobox, Nalgene®
Supplier: Thermo Fisher Scientific
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION
384-WELL GLASS MICROPLATE 74ΜL VOLUME P 1 * 1 Pce
Supplier: MARKET SOURCE PART PROCESS
Cell culture plates and Multidishes, Nunclon™ Sphera™
Supplier: Thermo Fisher Scientific
Insert System, BRAND<i>plates</i>®
Supplier: Brand
PCR plates, 96-well, segmented, Axygen®
Supplier: Corning
VESSEL 1L AMBER PLASTIC FOOTED 1 * 1 Pce
Supplier: MARKET SOURCE PART PROCESS