Competitive, selective EAAT2 inhibitor 1 * 10 mg
Catalog # MSPPHB0246-10MG
Supplier: MARKET SOURCE PART PROCESS
Competitive, selective EAAT2 inhibitor 1 * 10 mg
Catalog # MSPPHB0246-10MG
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR®, Conical Sample Containers, with Snap-On Lids
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection