U-PLEX NHP MIP-1β Assay, 384-well 1 * 25 items
Catalog # MESOK256UKK-4
Supplier: MESO SCALE DISCOVERY
U-PLEX NHP MIP-1β Assay, 384-well 1 * 25 items
Catalog # MESOK256UKK-4
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Water, PCR grade for PCR
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Decontamination solution, RNase AWAY®
Supplier: Thermo Fisher Scientific
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
E.Z.N.A.® Plasmid Mini kit I
Supplier: OMEGA BIO-TEK
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection