U-PLEX Human gp130 (soluble) Assay, SECTOR 1 * 5 items
Catalog # MESOK151P7K-2
Supplier: MESO SCALE DISCOVERY
U-PLEX Human gp130 (soluble) Assay, SECTOR 1 * 5 items
Catalog # MESOK151P7K-2
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Buffer CF (1000 mL) Lysis Buffer CF bottle of 1000 mL 1 * 1.000 mL
Supplier: MACHEREY-NAGEL
Savinase® 12 T 1 * 50 g
Supplier: Strem Chemicals, Inc.
HRP STREPTAVIDIN LIQ 1 * 1 mL
Supplier: SERACARE LIFE SCIENCES
R-PLEX Human Osteoactivin Assay 5 plate 1 * 1 KIT
Supplier: MESO SCALE DISCOVERY
R-PLEX HUMAN NEUROFILAMENT L ASSAY, SECTOR (5 PL) 1 * 1 KIT
Supplier: MESO SCALE DISCOVERY
R-PLEX Human SCF Assay, SECTOR 1 * 5 items
Supplier: MESO SCALE DISCOVERY
QuikChange II
Supplier: AGILENT
V-PLEX Plus Human PlGF Kit, SECTOR 1 * 1 items
Supplier: MESO SCALE DISCOVERY
V-PLEX COVID-19 Coronavirus Panel 2 (ACE2) Kit 1 * 5 items
Supplier: MESO SCALE DISCOVERY
EasySep™ Buffer
Supplier: STEMCELL Technologies
Agarose
Supplier: PanReac AppliChem
S-PLEX Human IL-9 Kit, QuickPlex 1 * 5 items
Supplier: MESO SCALE DISCOVERY