R-PLEX Human HSP27/HspB1 Assay, QuickPlex 1 * 5 items
MESOK151AGHR-22
Frequently Bought Together
Transfer pipettes
: RATIOLAB
Tissue DNA kit, genomic DNA isolation from tissue, E.Z.N.A.®
: OMEGA BIO-TEK
VWR® Serological Pipette, Standard Line
: VWR Collection
Fast DNA™ SPIN kit for soil, MP Biomedicals
: MP Biomedicals
VWR®, dNTPs
: VWR Chemicals
Decontamination solution, RNase AWAY®
: Thermo Fisher Scientific
Serological pipettes, Stripette™
: Corning
Peqlab peqGOLD, Agarose, Universal
: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Petri Dish, PS
: VWR Collection
Buffer CF (1000 mL) Lysis Buffer CF bottle of 1000 mL 1 * 1.000 mL
: MACHEREY-NAGEL
Mag-Bind® RXNPure Plus kit
: OMEGA BIO-TEK
RNA ScreenTape Analysis
: AGILENT