R-PLEX Human BMP-7 Assay, QuickPlex 1 * 5 items
Catalog # MESOK151D1R-22
Supplier: MESO SCALE DISCOVERY
R-PLEX Human BMP-7 Assay, QuickPlex 1 * 5 items
Catalog # MESOK151D1R-22
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
VWR®, Sampling Spoons
Supplier: VWR Collection
Decontamination solution, DNA AWAY®
Supplier: Thermo Fisher Scientific
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Decontamination solution, RNase AWAY®
Supplier: Thermo Fisher Scientific
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
E.Z.N.A.® Plasmid Mini kit I
Supplier: OMEGA BIO-TEK
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
HCCA matrix
Supplier: BRUKER DALTRONIK