U-PLEX NHP IL-1RA Assay, QuickPlex 1 * 5 items
MESOK156XPK-22
Frequently Bought Together
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
2-Propanol ≥99% for biotechnology
: VWR Chemicals
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
Decontamination solution, RNase AWAY®
: Thermo Fisher Scientific
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Kits for Precellys®
: Bertin Instruments
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Flow Cell Wash Kit
: Oxford Nanopore Technologies
Native barcoding auxiliary kit v14, Oxford Nanopore Technologies
: Oxford Nanopore Technologies
Ligation Sequencing Kits
: Oxford Nanopore Technologies
HCCA matrix
: BRUKER DALTRONIK