U-PLEX NHP I-TAC Assay, SECTOR 1 * 5 items
MESOK156UWK-2
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Decontamination solution, RNase AWAY®
: Thermo Fisher Scientific
VWR® Standard Line Transfer Pipettes
: VWR Collection
Decontamination solution, DNA AWAY®
: Thermo Fisher Scientific
VWR®, Sampling Spoons
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Centrifuge tubes, 15 ml, Corning®
: Corning
VWR® PCR Tubes, 0,2 ml
: VWR Collection