R-PLEX Human Elastase-2 Assay, QuickPlex 1 * 5 items
Catalog # MESOK151ABKR-22
Supplier: MESO SCALE DISCOVERY
R-PLEX Human Elastase-2 Assay, QuickPlex 1 * 5 items
Catalog # MESOK151ABKR-22
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Decontamination solution, DNA AWAY®
Supplier: Thermo Fisher Scientific
Decontamination solution, RNase AWAY®
Supplier: Thermo Fisher Scientific
Peqlab peqGOLD, Agarose, Universal
Supplier: VWR Chemicals
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
E.Z.N.A.® Plasmid Mini kit I
Supplier: OMEGA BIO-TEK
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection