R-PLEX Mouse IL-1α Assay, QuickPlex 1 * 5 items
MESOK1521YR-22
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Decontamination solution, RNase AWAY®
: Thermo Fisher Scientific
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR® PCR Tubes, 0,2 ml
: VWR Collection
Decontamination solution, DNA AWAY®
: Thermo Fisher Scientific
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
E.Z.N.A.® Plasmid Mini kit I
: OMEGA BIO-TEK
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection