R-PLEX Human Thyroglobulin Antibody Set 1 * 5 items
Catalog # MESOF21AGK-3
Supplier: MESO SCALE DISCOVERY
R-PLEX Human Thyroglobulin Antibody Set 1 * 5 items
Catalog # MESOF21AGK-3
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection