R-PLEX Human P-Selectin Antibody Set 1 * 5 items
Catalog # MESOF21ZM-3
Supplier: MESO SCALE DISCOVERY
R-PLEX Human P-Selectin Antibody Set 1 * 5 items
Catalog # MESOF21ZM-3
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Transfer pipettes
Supplier: RATIOLAB
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Cell culture dishes, Falcon®
Supplier: Corning
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Deep well storage plates, 96-well, 0,8 ml
Supplier: THERMO MATRIX TECHNOLOGIES
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Aspirating pipettes, Falcon®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR®, Serological Pipettes, Open End
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, Sterile
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VIAL 55ML CUSTODY CLEAR 19 1 * 450 items
Supplier: Capitol Europe