R-PLEX Human CD44 Antibody Set 1 * 5 items
Catalog # MESOF21AJH-3
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Stoppers, press down, VacuCap®
Supplier: Simport Scientific
Cryoboxes, 133×133 mm
Supplier: TENAK
VWR® Cell Culture Dishes
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR®, High Performance Blender Bags
Supplier: VWR Collection
Pipette tray for drawer
Supplier: KARTELL
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryo coder cap inserts
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Microcentrifuge tubes, 1,5 ml, Secure-Lock™ cap
Supplier: Simport Scientific