MSD Blocker D-G 1 * 1 mL
MESOR93BH-3
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
: VWR Chemicals
Kjeldahl tablets
: Merck
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Potassium hydroxide 85.0-100.5%, pellets Ph. Eur.
: VWR Chemicals