U-PLEX Mouse MCP-1 Antibody Set 1 * 1 items
Catalog # MESOB22UG-2
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Cryoboxes, 133×133 mm
Supplier: TENAK
Cell culture flasks, Falcon®
Supplier: Corning
Strips of 8 PCR tubes with attached flat individual caps
Supplier: Brand
Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Supplier: Corning
VWR®, Cryo coder cap inserts
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
MSD GOLD 96-WELL STREPTAVIDIN SECTOR PLATE PACK 1 * 5 items
Supplier: MESO SCALE DISCOVERY
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Supplier: Simport Scientific
Culture tubes, sterile, Cultubes™
Supplier: Simport Scientific
VWR®, Cell Culture Insert Plates, PC
Supplier: VWR Collection
µ-PLATE 96 WELL ROUND 1 * 1 Pce
Supplier: MARKET SOURCE PART PROCESS
EASYSTRAINER SMALL SAMPLE PACK, ONE OF EACH MESH SIZE 1 * 1 items
Supplier: Greiner Bio-One
Sample tubes
Supplier: Simport Scientific