MSD Blocker D-M 1 * 0,2 mL
Catalog # MESOR93BM-1
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
EOSIN RAL 555 1L 1 * 1 L
Supplier: RAL
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Kjeldahl catalyst acc. to Wieninger
Supplier: Merck
Eosin Y (yellowish) in aqueous solution, Q Path® for microscopy
Supplier: VWR Chemicals
Ammonia solution 20%, GPR RECTAPUR®
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
TAMPON PH 2 SELON NORME NF T 90-431 ( a autoclaver ) 1 * 1 L
Supplier: PROLABO CMP
NANOCOLOR® Photometric Tube Tests, Nitrate
Supplier: MACHEREY-NAGEL
Immersion oil
Supplier: RAL
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
ISOPROPANOL 30% 1 * 2,5 L
Supplier: PROLABO CMP
Polyester-wax for microscopy
Supplier: VWR Chemicals
ALKALINE-CYANIDE REAGENT 1 * 500 mL
Supplier: Hach