Orders LinkContactUsLinkComponent
Label Sheets, Core2Write
Label Sheets, Core2Write
Core2Write label, sterile, irradiated, White, 101,6×76,2 mm, 6 labels/sheet, 500 sheets/case
Catalog # VELTC2WR4X3SHEET02
Supplier:   VELTEK ASSOCIATES
Label Sheets, Core2Write
Catalog # VELTC2WR4X3SHEET02
Supplier:  VELTEK ASSOCIATES
Label Sheets, Core2Write
Label Sheets, Core2Write
Core2Write label, sterile, irradiated, White, 101,6×76,2 mm, 6 labels/sheet, 500 sheets/case
Price
4 800,00 €
Log in for availability
1 * 3.000 ST
Product Details & Documents
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures.
  • Can be applied to sub-zero surfaces
  • Perforation between labels is available
All labels can be customised or left blank.
Ordering information: Also available in sheets (sold separately).
Specifications
  • Colour:
    White
  • Description:
    Core2Write label, sterile, irradiated
  • Dimensions:
    101,6×76,2 mm
  • Seq:
    05
  • Packed:
    6 labels/sheet, 500 sheets/case