Orders LinkContactUsLinkComponent
Label Sheets, Core2Write
Core2Write label, sterile, irradiated, White, 101,6×50,8 mm, 10 labels/sheet, 500 sheets/case
  VELTC2WR4X2SHEET02
Label Sheets, Core2Write
  VELTC2WR4X2SHEET02
Label Sheets, Core2Write
Label Sheets, Core2Write
Core2Write label, sterile, irradiated, White, 101,6×50,8 mm, 10 labels/sheet, 500 sheets/case
4 800,00 €
1 * 5.000 ST
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures.
  • Can be applied to sub-zero surfaces
  • Perforation between labels is available
All labels can be customised or left blank.
 : Also available in sheets (sold separately).
  • Colour:
    White
  • Description:
    Core2Write label, sterile, irradiated
  • Dimensions:
    101,6×50,8 mm
  • Seq:
    04
  • Packed:
    10 labels/sheet, 500 sheets/case