1 COMPONENT: HG 0.25MG/L IN HCL 5% 1 * 250 mL
Catalog # CMPLRM076383L25
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
1 COMPONENT: HG 0.25MG/L IN HCL 5% 1 * 250 mL
Catalog # CMPLRM076383L25
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Papanicolaou's solution 2a Orange G (OG 6), Sigma-Aldrich®
Supplier: Merck
Test strips, molybdenum, MQuant®, Supelco®
Supplier: Merck
Test papers, lead acetate papers
Supplier: MACHEREY-NAGEL
L(+)-Histidine monohydrochloride monohydrate 98.5-101.0% (dried basis), EMPROVE® EXPERT Ph. Eur., BP, JP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Colorimetric rapid tests, phosphate, VISOCOLOR®
Supplier: MACHEREY-NAGEL
Calcium hydroxide ≥96%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium iodide 10% in aqueous solution
Supplier: PROLABO CMP
Ammonia solution 20%, GPR RECTAPUR®
Supplier: VWR Chemicals
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
METHYLENE BLUE RAL 555 AQUEOUS SOL. 1L 1 * 1 L
Supplier: RAL
ACIDE CHLORHYDRIQUE 4.4M 1 * 10 L
Supplier: PROLABO CMP
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
POTASSIUM BROMATE KBR/KBRO3 0.5 MOL/L 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
ALKALINE-CYANIDE REAGENT 1 * 500 mL
Supplier: Hach