COMPONENTS 13 MIX 1 * 100 mL
CMPLRM110120L1
New Product
Frequently Bought Together
pH indicator test strips, PEHANON®
: MACHEREY-NAGEL
Sodium hydroxide 97.0-100.5%, pellets NF, JP, Ph. Eur., BP
: VWR Chemicals
MODIFIER-NH4H2PO4 MATRIX 1 * 1 items
: PerkinElmer
Test strips, water hardness, AQUADUR®
: MACHEREY-NAGEL
Formaldehyde 37% (w/w) in aqueous solution stabilised ACS
: Thermo Fisher Scientific
Citric acid
: Hach
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
: CUSTOM MADE CHEMICALS