pH Buffers
pH buffer, 3,9 M NaCl, 4,010, 6×250 ml, 1.500 mL
Catalog # METN52004104
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1.500 mL
- Description:pH buffer, 3,9 M NaCl
- Pk Info:6×250 ml
- pH value:4,010



