ContactUsLinkComponent
A unique combination of PCR products and a number of proprietary plasmids digested with appropriate restriction enzymes. Phenol extr acted and equilibrated to 10 mM Tris-HCl (pH 8.0) and 10 mM EDTA. Covers the size range 100-10,000 base pairs. Number 1 * 500 µl
Catalog # ANTIA318972-500
Supplier:  ANTIBODIES.COM
New Product
A unique combination of PCR products and a number of proprietary plasmids digested with appropriate restriction enzymes. Phenol extr acted and equilibrated to 10 mM Tris-HCl (pH 8.0) and 10 mM EDTA. Covers the size range 100-10,000 base pairs. Number 1 * 500 µl
Catalog # ANTIA318972-500
Supplier:  ANTIBODIES.COM