ContactUsLinkComponent
Microscope cover glasses, with works certificate, 24x24 mm, borosilicate glass D 263® M, thickness No. 1.5H (0.17±0.005 mm), CE marked 200 pcs./hinged-lid plastic box 1 * 2.000 items
Catalog # MARI0107062091
Supplier:  MARIENFELD
New Product
Microscope cover glasses, with works certificate, 24x24 mm, borosilicate glass D 263® M, thickness No. 1.5H (0.17±0.005 mm), CE marked 200 pcs./hinged-lid plastic box 1 * 2.000 items
Catalog # MARI0107062091
Supplier:  MARIENFELD