ContactUsLinkComponent
This innovative, split-frame Switch-Grid rack holds up to two different size tubes in the same rack and allows users to quickly switch grids to hold other sizes as needed. Each rack frame includes 5 sets of grids in varying size. 1 * 1 items
Catalog # BELAF18745-1000
New Product
This innovative, split-frame Switch-Grid rack holds up to two different size tubes in the same rack and allows users to quickly switch grids to hold other sizes as needed. Each rack frame includes 5 sets of grids in varying size. 1 * 1 items
Catalog # BELAF18745-1000