Orders LinkContactUsLinkComponent
6-VIC phosphoramidite
100 µmol
Catalog # AATB6087
Supplier:   AAT Bioquest
6-VIC phosphoramidite
Catalog # AATB6087
Supplier:  AAT Bioquest
CAS Number:  
6-VIC phosphoramidite
100 µmol
CAS Number:  
Price
486,00 €
Log in for availability
1 * 1 EA
Product Details & Documents
6-VIC phosphoramidite is an asymmetrical xanthene dye widely used to prepare fluorescence-labeled oligonucleotides and design qPCR probes, such as TaqMan, Molecular Beacon, and Scorpion.
  • Stable for 6 months
Quality tested in-house
 : For Research Use Only
Specifications
Specifications
  • Pk:
    100 µmol