RNA ELUTION BUFFER 500ML 1 * 500 mL
Catalog # OMEGRNAELUTION-500
Supplier: OMEGA BIO-TEK
New Product
Frequently Bought Together
Filter tips, uTIP™
Supplier: BIOTIX
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
VWR®, Microscope Slides
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Adhesive PCR film and foil seals
Supplier: Thermo Fisher Scientific
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF