SYSTEM KIT CREATININE FS 8X50ML 1 * 1 KIT
Catalog # DIAS117119910704
Supplier: DIAGNOSTIC SYSTEMS DIASYS
New Product
SYSTEM KIT CREATININE FS 8X50ML 1 * 1 KIT
Catalog # DIAS117119910704
Supplier: DIAGNOSTIC SYSTEMS DIASYS
New Product
Frequently Bought Together
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
pH indicator test strips, PEHANON®
Supplier: MACHEREY-NAGEL
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
GLUTAMINE V2 BIO HT
Supplier: ROCHE
Test strips, peracetic acid, Dosatest
Supplier: VWR Chemicals
ELECTROFERICYAN SOLUTION 1 * 5 L
Supplier: PROLABO CMP
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
VWR® Rota® Duo, pH indicator papers
Supplier: VWR Chemicals
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Periodic acid solution, Q Path® for microscopy
Supplier: VWR Chemicals
Reagents for ammonia determination using Lovibond® MD photometers
Supplier: Lovibond
BOLA-VACUUM TRAP 1 * 1 items
Supplier: Bohlender
ALKALINE-CYANIDE REAGENT 1 * 500 mL
Supplier: Hach
Test kits, photometric, Nitrite, LCK cuvette tests
Supplier: Hach