Order Entry
Belgium
Orders LinkContactUsLinkComponent
M-Aprons, 0,5 mm

M-Aprons, 0,5 mm

M-Aprons 0,5 mm, 900×1200 mm, Black
Catalog # BSNC090-125
Supplier:   Basan
M-Aprons, 0,5 mm
Catalog # BSNC090-125
Supplier:  Basan
M-Aprons, 0,5 mm

M-Aprons 0,5 mm, 900×1200 mm, Black

Price
55,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

M-Aprons can be utilised in various fields, such as the chemical industry, healthcare, food processing, and cleaning companies.

All aprons are machine washable up to 60 °C but should not be dry cleaned. They are not suitable for use with heat or open flames. Store them at room temperature and avoid creases by rolling them up or hanging them by the neck straps.

Specifications
  • Colour:
    Black
  • Description:
    M-Aprons 0,5 mm, 900×1200 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.