ContactUsLinkComponent
DIN 12781 enclosed form, upper part length: 250 mm, diameter: 11 ± 0.5 mm, measuring range: -100+30°C in 1°C, capillary: uncoated prismatic, red toluene filling, lower part length: 300 mm, diameter: 8 ± 0.5 mm, immersion depth: 280 mm, total length: 1 * 1 items
Catalog # SCNL1052706
New Product
DIN 12781 enclosed form, upper part length: 250 mm, diameter: 11 ± 0.5 mm, measuring range: -100+30°C in 1°C, capillary: uncoated prismatic, red toluene filling, lower part length: 300 mm, diameter: 8 ± 0.5 mm, immersion depth: 280 mm, total length: 1 * 1 items
Catalog # SCNL1052706