ContactUsLinkComponent
EAU HIPERSOLV CHROMANORM HPLC,, VWRC23595.**,, 1248,75,,mL ACIDE ORTHOPHOSPHORIQUE 85 % HIPERSOLV C,, VWRC153154D,, 1,25,,mL METHANOL HIPERSOLV HPLC ISOCRATIC GRADE,, VWRC20837.**,, 1250,,mL 1 * 2,5 L
Catalog # PRLS90326.320
Supplier:  PROLABO CMP
New Product
EAU HIPERSOLV CHROMANORM HPLC,, VWRC23595.**,, 1248,75,,mL ACIDE ORTHOPHOSPHORIQUE 85 % HIPERSOLV C,, VWRC153154D,, 1,25,,mL METHANOL HIPERSOLV HPLC ISOCRATIC GRADE,, VWRC20837.**,, 1250,,mL 1 * 2,5 L
Catalog # PRLS90326.320
Supplier:  PROLABO CMP