Réactif DINO 975 ml Eau 56 g Acide oxalique dihydraté 10 ml Acide fluorhydrique 48% 14 ml Acide sulfurique 96% 1 * 1 L
Catalog # PRLS90326.290
Supplier: PROLABO CMP
Réactif DINO 975 ml Eau 56 g Acide oxalique dihydraté 10 ml Acide fluorhydrique 48% 14 ml Acide sulfurique 96% 1 * 1 L
Catalog # PRLS90326.290
Supplier: PROLABO CMP
Frequently Bought Together
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Joint grease, K.W.S.
Supplier: POLLATH
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Immersion oil for microscopy, Sigma-Aldrich®
Supplier: Merck
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
1-Heptanesulphonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS