20 COMPONENTS MIX 1 * 200 mL
Catalog # CMPLRM073578L2
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
pH buffers, ready to use
Supplier: WTW
Buffer solutions
Supplier: Hanna
UV/Visible standards, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Humidity standards
Supplier: ROTRONIC INSTRUMENTS
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals