Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Water Separator

Water separator, two way stopcock, NS 18,8/2,5 mm, 10 ml, 0,1 ml, 29/32 NS, 29/32 NS
  NEUB03829010292919
Water Separator
  NEUB03829010292919
Water Separator

Water separator, two way stopcock, NS 18,8/2,5 mm, 10 ml, 0,1 ml, 29/32 NS, 29/32 NS

 
518,00 €
   
1 * 1 ST
 

Water separator made of Borosilicate glass 3.3.

  • Graduation 10 ml in 1/10, 0-point on the stopcock
  • With 2-way tap
 
  • Cone to condenser:
    29/32 NS
  • Cone to flask:
    29/32 NS
  • Capacity:
    10 ml
  • Description:
    Water separator, two way stopcock, NS 18,8/2,5 mm
  • Seq:
    04
  • Graduation:
    0,1 ml