Decoquinate 98%
Glass bottle, 5 g
N° de catalogue 472090050.
Fournisseur: Thermo Fisher Scientific
Decoquinate 98%
N° de catalogue 472090050.
Fournisseur: Thermo Fisher Scientific
Numéro CAS: 18507-89-6
Jump to:
Product Details & Documents
Synonyms: Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester
- CAS Number:18507-89-6
- Formula:C₂₄H₃₅NO₅
- MDL Number:MFCD00673686
- Molecular Weight:417.55 g/mol
- Merck Index:13,02874
- Storage Temperature:Ambient
- EINECS:242-389-1
Specifications
Specification Test Results
- Appearance:white to light yellow to light beige crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Melting point:241°C to 245°C
- Water:≤1 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



