Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Decoquinate 98%

Glass bottle, 1 g
Catalog # 472090010.
Decoquinate 98%
Catalog # 472090010.
CAS Number:  18507-89-6
Decoquinate 98%

Glass bottle, 1 g

CAS Number:  18507-89-6
Price
69,70 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester
  • CAS Number:18507-89-6
  • Formula:C₂₄H₃₅NO₅
  • MDL Number:MFCD00673686
  • Molecular Weight:417.55 g/mol
  • Merck Index:13,02874
  • Storage Temperature:Ambient
  • EINECS:242-389-1
Specifications

Specification Test Results

  • Appearance:
    white to light yellow to light beige crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %
  • Melting point:
    241°C to 245°C
  • Water:
    ≤1 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications