Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multi-Angle Light-Scattering Detector, LenS3™

Multi-Angle Light-Scattering Detector, LenS3™

LenS3™ Multi-Angle Light-Scattering detector
Catalog # TOYO0040000
Supplier:   TOSOH Bioscience
Multi-Angle Light-Scattering Detector, LenS3™
Catalog # TOYO0040000
Supplier:  TOSOH Bioscience
Multi-Angle Light-Scattering Detector, LenS3™

LenS3™ Multi-Angle Light-Scattering detector

Price
73 210,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Tosoh’s LenS3™ Multi-Angle Light-Scattering detector combines high-sensitivity MW and Rg determinations with flexibility and ease of use.

  • Characterisation of molecules down to 200 to 10⁷ Da and Rg down to <2 to >300 nm
  • Optics design for low noise and high scattering intensity
  • Compatible with GPC, SEC, HPLC and UHPLC systems
  • Compatible with all common solvents (THF, HFIP, aqueous salt buffers)
  • Modular software for simultaneous acquisition data analysis, sequence editing
  • Analysis modules for step-by-step guidance
  • Proprietary conical flow path
  • Laser: Diode, 20 mW ±5, wavelength 505 nm
  • Baseline noise: <1 mV (in THF at 1 ml/min)
  • Wetted materials: Teflon, PEEK, glass, stainless steel

The LenS3™ Multi-Angle Light-Scattering detector enables you to directly measure the molecular weight and radius of gyration of polymers, polysaccharides, and biopolymers such as proteins or delivery particles. Its high sensitivity allows the characterisation of low sample amounts, molecules with low MW or determination of Rg values downt to <10 nm. It serves you in combination with any SEC device such as GPC, HPLC or UHPLC instruments.

Maximum flow rate: 2 ml/min
Acquisition rate: 10 Hz
Dimensions (W×D×H): 36,5×48,5×13 mm
Weight: 16 kg

Specifications
  • Description:
    LenS3™ Multi-Angle Light-Scattering detector
  • Seq:
    01
Services

Product Information

  • Catalog #
    Description
    Availability
    Price
  • TOYO0046040
    Operational Qualification (OQ)
    Log in for availability
    1 * 1 ST - 3 750,00 €

  • TOYO0046039
    Installation Qualification (IQ)
    Log in for availability
    1 * 1 ST - 1 880,00 €

  • TOYO0046038
    Installation
    Log in for availability
    1 * 1 ST - 3 750,00 €

Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Stainless steel inline filter housing, Int.Ø: 13 mm
    Stainless steel inline filter housing, Int.Ø: 13 mm
    TOYO0040002
    Log in for availability
    1 * 1 ST - 143,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Post column filter 0,2 µm, 13 mm nylon
    Post column filter 0,2 µm, 13 mm nylon
    TOYO0040032
    Log in for availability
    1 * 100 ST - 13,10 €

Product Information

  • Catalog #
    Availability
    Price
  • Post pump filter 0,03 µm, 13 mm PES
    Post pump filter 0,03 µm, 13 mm PES
    TOYO0040048
    Log in for availability
    1 * 100 ST - 151,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Post column filter 0,1 µm, 13 mm PTFE
    Post column filter 0,1 µm, 13 mm PTFE
    TOYO0040031
    Log in for availability
    1 * 100 ST - 13,10 €

Product Information

  • Catalog #
    Availability
    Price
  • Post column filter 0,2 µm, 13 mm PES
    Post column filter 0,2 µm, 13 mm PES
    TOYO0040049
    Log in for availability
    1 * 100 ST - 151,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Post column filter 0,2 µm, 13 mm PTFE
    Post column filter 0,2 µm, 13 mm PTFE
    TOYO0040074
    Log in for availability
    1 * 100 ST - 38,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Stainless steel frits, Int.Ø: 13 mm (for inline filter housing)
    Stainless steel frits, Int.Ø: 13 mm (for inline filter housing)
    TOYO0040030
    Log in for availability
    1 * 10 ST - 60,50 €

Product Information

  • Catalog #
    Availability
    Price
  • UHPLC stainless steel inline micro filter
    UHPLC stainless steel inline micro filter
    TOYO0040043
    Log in for availability
    1 * 5 ST - 303,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Post pump filter 0,1 µm, 13 mm nylon
    Post pump filter 0,1 µm, 13 mm nylon
    TOYO0040042
    Log in for availability
    1 * 100 ST - 13,10 €