Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Water Separator

Water separator, tap NS 18,8/2,5 mm, 10 ml, 0,1 ml, 29/32 NS, 29/32 NS
  NEUB-90-10292919
Water Separator
  NEUB-90-10292919
Water Separator

Water separator, tap NS 18,8/2,5 mm, 10 ml, 0,1 ml, 29/32 NS, 29/32 NS

 
528,00 €
   
1 * 1 ST
 

Water separator made of Borosilicate glass 3.3.

  • Graduation 10 ml in 1/10, 0-point on the stopcock
  • With 2-way tap
 
  • Cone to condenser:
    29/32 NS
  • Cone to flask:
    29/32 NS
  • Capacity:
    10 ml
  • Description:
    Water separator, tap NS 18,8/2,5 mm
  • Seq:
    02
  • Graduation:
    0,1 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.