4 components: K 3mg/l Na 10mg/l Ca 20mg/l Mg 8mg/l in HNO3 2% 1 * 1.000 mL
CMPLRM0690341L.3
4 components: K 3mg/l Na 10mg/l Ca 20mg/l Mg 8mg/l in HNO3 2% 1 * 1.000 mL
CMPLRM0690341L.3
Frequently Bought Together
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Sodium hydroxide 50% in aqueous solution, TECHNICAL
: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Sodium hydroxide ≥97.0%, shot, TECHNICAL
: VWR Chemicals
Calcium hydroxide 95%
: Thermo Fisher Scientific
Zinc protoporphyrin guanylate cyclase inhibitor, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Fast Red RC Salt, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY